4,5-Difluor-2-nitrobenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS20317G100 AS20317G25
ACTIAS20317G100EA 177 CHF
ACTIAS20317G100 ACTIAS20317G25
4,5-Difluor-2-nitrobenzoesäure
4,5-Difluor-2-nitrobenzoesäure
Formel: C₇H₃F₂NO₄
MW: 203,1 g/mol
MDL Number: MFCD00799521
CAS-Nummer: 20372-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR