2-Fluor-6-nitrobenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Fluoro-2-nitrobenzoic acid

AS20463G25 AS20463G5
ACTIAS20463G25EA 76 CHF
ACTIAS20463G25 ACTIAS20463G5
2-Fluor-6-nitrobenzoesäure
2-Fluoro-6-nitrobenzoic acid
Formel: C₇H₄FNO₄
MW: 185,11 g/mol
MDL Number: MFCD01862079
CAS-Nummer: 385-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR