Methyl-5-fluor-2-nitrobenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 5-fluoro-2-nitrobenzoate, 5-Fluorobenzoic acid methyl ester

AS23571G25 AS23571G5
ACTIAS23571G25EA 114 CHF
ACTIAS23571G25 ACTIAS23571G5
Methyl-5-fluor-2-nitrobenzoat
Methyl-5-fluor-2-nitrobenzoat
Formel: C₈H₆FNO₄
MW: 199,14 g/mol
MDL Number: MFCD03425619
CAS-Nummer: 393-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR