2-Fluorbenzamidin Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluorobenzimidamide hydrochloride, 2-Fluorophenylamidine hydrochloride, 2-Fluorobenzene-1-carboximidamide hydrochloride

AS30170G1 AS30170G5
ACTIAS30170G1EA 0 CHF
ACTIAS30170G1 ACTIAS30170G5
2-Fluorbenzamidin Hydrochlorid
2-Fluorbenzamidin Hydrochlorid
Formel: C₇H₈ClFN₂
MW: 174,61 g/mol
MDL Number: MFCD04114435
CAS-Nummer: 57075-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR