4-Fluor-3-hydroxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS35107G1 AS35107G5
ACTIAS35107G1EA 0 CHF
ACTIAS35107G1 ACTIAS35107G5
4-Fluor-3-hydroxybenzonitril
4-Fluor-3-hydroxybenzonitril
Formel: C₇H₄FNO
MW: 137,11 g/mol
MDL Number: MFCD08689519
CAS-Nummer: 186590-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR