2,6-Difluor-4-hydroxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Cyano-3,5-difluorophenol

AS20513G1 AS20513G5
ACTIAS20513G1EA 0 CHF
ACTIAS20513G1 ACTIAS20513G5
2,6-Difluor-4-hydroxybenzonitril
2,6-Difluor-4-hydroxybenzonitril
Formel: C₇H₃F₂NO
MW: 155.1 g/mol
MDL Number: MFCD03094499
CAS-Nummer: 123843-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR