2,3-Difluor-4-hydroxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS33687G1 AS33687G5
ACTIAS33687G1EA 0 CHF
ACTIAS33687G1 ACTIAS33687G5
2,3-Difluor-4-hydroxybenzonitril
2,3-Difluor-4-hydroxybenzonitril
Formel: C₇H₃F₂NO
MDL Number: MFCD00269596
CAS-Nummer: 126162-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR