4-Fluor-3-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS22531G100 AS22531G25
ACTIAS22531G100EA 0 CHF
ACTIAS22531G100 ACTIAS22531G25
4-Fluor-3-methoxybenzonitril
4-Fluor-3-methoxybenzonitril
Formel: C₈H₆FNO
MDL Number: MFCD01569371
CAS-Nummer: 243128-37-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR