4-Fluor-2-methoxybenzonitril
Lieferant: ACTIVATE SCIENTIFIC
AS35189G1
AS35189G10
AS35189G5
ACTIAS35189G1EA
0
CHF
ACTIAS35189G1
ACTIAS35189G10
ACTIAS35189G5
4-Fluor-2-methoxybenzonitril
4-Fluor-2-methoxybenzonitril
Formel:
C₈H₆FNO MW: 151.14 g/mol |
MDL Number:
MFCD04116335 CAS-Nummer: 191014-55-8 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...