3-Fluor-2-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS35096G10 AS35096G5
ACTIAS35096G10EA 0 CHF
ACTIAS35096G10 ACTIAS35096G5
3-Fluor-2-methoxybenzonitril
3-Fluor-2-methoxybenzonitril
Formel: C₈H₆FNO
MDL Number: MFCD06637312
CAS-Nummer: 77801-22-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR