4,5-Difluor-2-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS36742G1 AS36742G5
ACTIAS36742G1EA 0 CHF
ACTIAS36742G1 ACTIAS36742G5
4,5-Difluor-2-methoxybenzonitril
4,5-Difluor-2-methoxybenzonitril
Formel: C₈H₅F₂NO
MDL Number: MFCD06660346
CAS-Nummer: 425702-28-9
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR