2-Fluorisonicotinonitril
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Cyano-2-fluoropyridine
AS51292G1
AS51292G5
ACTIAS51292G1EA
0
CHF
ACTIAS51292G1
ACTIAS51292G5
2-Fluorisonicotinonitril
2-Fluorisonicotinonitril
Formel:
C₆H₃FN₂ |
MDL Number:
MFCD09027299 CAS-Nummer: 3939-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...