3,5-Difluor-4-hydroxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS35871G1 AS35871G5
ACTIAS35871G1EA 0 CHF
ACTIAS35871G1 ACTIAS35871G5
3,5-Difluor-4-hydroxybenzaldehyd
3,5-Difluoro-4-hydroxybenzaldehyde
Formel: C₇H₄F₂O₂
MDL Number: MFCD06797917
CAS-Nummer: 118276-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR