2,6-Difluor-4-hydroxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS34365G1 AS34365G5
ACTIAS34365G1EA 0 CHF
ACTIAS34365G1 ACTIAS34365G5
2,6-Difluor-4-hydroxybenzaldehyd
2,6-Difluoro-4-hydroxybenzaldehyde
Formel: C₇H₄F₂O₂
MDL Number: MFCD07772042
CAS-Nummer: 532967-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR