5-Fluor-o-anisaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Fluoro-2-anisaldehyde, 5-Fluoro-2-methoxybenzaldehyde

AS32301G1 AS32301G10 AS32301G5
ACTIAS32301G1EA 0 CHF
ACTIAS32301G1 ACTIAS32301G10 ACTIAS32301G5
5-Fluor-o-anisaldehyd
5-Fluor-o-anisaldehyd
Formel: C₈H₇FO₂
MDL Number: MFCD00143458
CAS-Nummer: 19415-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR