3-Fluor-2-methylbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Fluoro-o-tolualdehyde, 3-Fluoro-2-tolualdehyde, 3-Fluoro-o-tolylaldehyde

AS20640G25 AS20640G5
ACTIAS20640G25EA 119 CHF
ACTIAS20640G25 ACTIAS20640G5
3-Fluor-2-methylbenzaldehyd
3-Fluoro-2-methylbenzaldehyde
Formel: C₈H₇FO
MW: 138,14 g/mol
MDL Number: MFCD00075256
CAS-Nummer: 147624-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR