2-Fluor-5-methylbenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Fluoro-5-tolualdehyde, 6-Fluoro-m-tolylaldehyde, 6-Fluoro-m-tolualdehyde
AS31868G1
AS31868G10
AS31868G5
ACTIAS31868G1EA
0
CHF
ACTIAS31868G1
ACTIAS31868G10
ACTIAS31868G5
2-Fluor-5-methylbenzaldehyd
2-Fluoro-5-methylbenzaldehyde
Formel:
C₈H₇FO MW: 138,14 g/mol |
MDL Number:
MFCD02093762 CAS-Nummer: 93249-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...