4-(Hydroxymethyl)benzoesäure

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: HMBA4-Carboxybenzylalkohol

ACTIAS20975G25EA 61 CHF
ACTIAS20975G5 ACTIAS20975G25
4-(Hydroxymethyl)benzoesäure
4-(Hydroxymethyl)benzoic acid
Formel: C₈H₈O₃
MW: 152,15 g/mol
MDL Number: MFCD00017598
CAS-Nummer: 3006-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR