(2-Pyridyl)essigsäureethylester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 2-Pyridylacetate, Ethyl 2-pyridineacetate, Ethyl (2-pyridyl)acetate

AS20037G25 AS20037G5
ACTIAS20037G25EA 0 CHF
ACTIAS20037G25 ACTIAS20037G5
(2-Pyridyl)essigsäureethylester
(2-Pyridyl)essigsäureethylester
Formel: C₉H₁₁NO₂
Siedepunkt: 302 °C (1013 hPa)
Melting Pt: 86…90 °C
Dichte: 1,08 g/cm³ (20 °C)
MDL Number: MFCD00006359
CAS-Nummer: 2739-98-2
EINECS: 220-365-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR