4,5-Dicyan-1-methylimidazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4,5-Dicyano-1-methyl-1H-imidazole, 1-Methyl-1H-imidazole-4,5-dicarbonitrile, 1-Methylimidazole-4,5-dicarbonitrile

AS21869G100 AS21869G25 AS21869G5
ACTIAS21869G100EA 358 CHF
ACTIAS21869G100 ACTIAS21869G25 ACTIAS21869G5
4,5-Dicyan-1-methylimidazol
4,5-Dicyano-1-methylimidazole
Formel: C₆H₄N₄
MW: 132,12 g/mol
MDL Number: MFCD01567299
CAS-Nummer: 19485-35-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR