3-Amino-4-pyridincarbonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Amino-4-cyanopyridine

AS22160G1 AS22160G5
ACTIAS22160G1EA 124 CHF
ACTIAS22160G1 ACTIAS22160G5
3-Amino-4-pyridincarbonitril
3-Amino-4-pyridincarbonitril
Formel: C₆H₅N₃
MW: 119,13 g/mol
MDL Number: MFCD04035596
CAS-Nummer: 78790-79-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR