2-Chlor-4-methyl-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-3-nitro-4-picoline

AS7047G1 AS7047G5
ACTIAS7047G1EA 0 CHF
ACTIAS7047G1 ACTIAS7047G5
2-Chlor-4-methyl-3-nitropyridin
2-Chloro-4-methyl-3-nitropyridine
Formel: C₆H₅ClN₂O₂
MW: 172,57 g/mol
MDL Number: MFCD00012347
CAS-Nummer: 23056-39-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR