2-Chlor-5-fluor-4-iodpyridin
Lieferant: ACTIVATE SCIENTIFIC
AS32500G1
AS32500G5
ACTIAS32500G1EA
0
CHF
ACTIAS32500G1
ACTIAS32500G5
2-Chlor-5-fluor-4-iodpyridin
2-Chloro-5-fluoro-4-iodopyridine
Formel:
C₅H₂ClFIN MW: 257,43 g/mol |
MDL Number:
MFCD03095306 CAS-Nummer: 884494-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...