2-Chlor-3-fluor-4-iodpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS35823G1 AS35823G5
ACTIAS35823G1EA 30 CHF
ACTIAS35823G1 ACTIAS35823G5
2-Chlor-3-fluor-4-iodpyridin
2-Chloro-3-fluoro-4-iodopyridine
Formel: C₅H₂ClFIN
MW: 257,43 g/mol
MDL Number: MFCD04972389
CAS-Nummer: 148639-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR