Methyl-2-chlor-4-fluorbenzoat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms: Methyl 2-chloro-4-fluorobenzenecarboxylate, Methyl 2-chloro-4-fluorobenzoate, 2-Chloro-4-fluorobenzoic acid methyl ester
AS22671G100 AS22671G25
ACTIAS22671G100EA 172 CHF
ACTIAS22671G100 ACTIAS22671G25
Methyl-2-chlor-4-fluorbenzoat
Methyl-2-chlor-4-fluorbenzoat
Formel: C₈H₆ClFO₂
MW: 188,59 g/mol
MDL Number: MFCD00173937
CAS-Nummer: 85953-29-3