4'-Chlor-2'-fluoracetophenon

Lieferant: ACTIVATE SCIENTIFIC

AS34291G1 AS34291G5
ACTIAS34291G1EA 0 CHF
ACTIAS34291G1 ACTIAS34291G5
4'-Chlor-2'-fluoracetophenon
4'-Chlor-2'-fluoracetophenon
Formel: C₈H₆ClFO
MW: 172.59 g/mol
MDL Number: MFCD04116328
CAS-Nummer: 175711-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR