3-Chlor-5-fluorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS33224G1 AS33224G10 AS33224G5
ACTIAS33224G1EA 0 CHF
ACTIAS33224G1 ACTIAS33224G10 ACTIAS33224G5
3-Chlor-5-fluorbenzonitril
3-Chlor-5-fluorbenzonitril
Formel: C₇H₃ClFN
MW: 155.56 g/mol
MDL Number: MFCD03407967
CAS-Nummer: 327056-73-5
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR