2-Chlor-3-fluorbenzonitril
Lieferant: ACTIVATE SCIENTIFIC
AS36039G1
AS36039G5
ACTIAS36039G1EA
0
CHF
ACTIAS36039G1
ACTIAS36039G5
2-Chlor-3-fluorbenzonitril
2-Chlor-3-fluorbenzonitril
Formel:
C₇H₃ClFN MW: 155,56 g/mol |
MDL Number:
MFCD03407966 CAS-Nummer: 874781-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...