5-Chlor-2-fluor-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Chloro-2-fluoro-3-picoline

AS31514G1 AS31514G10 AS31514G5
ACTIAS31514G1EA 0 CHF
ACTIAS31514G1 ACTIAS31514G10 ACTIAS31514G5
5-Chlor-2-fluor-3-methylpyridin
5-Chlor-2-fluor-3-methylpyridin
Formel: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD03095094
CAS-Nummer: 375368-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR