2-Chlor-5-fluor-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-5-fluoro-3-picoline

AS33577G1 AS33577G5
ACTIAS33577G1EA 0 CHF
ACTIAS33577G1 ACTIAS33577G5
2-Chlor-5-fluor-3-methylpyridin
2-Chloro-5-fluoro-3-methylpyridine
Formel: C₆H₅ClFN
MW: 145,56 g/mol
MDL Number: MFCD04972725
CAS-Nummer: 38186-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR