3-Chlor-4-nitrobenzoesäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS33430G10 AS33430G100 AS33430G5
ACTIAS33430G10EA 0 CHF
ACTIAS33430G10 ACTIAS33430G100 ACTIAS33430G5
3-Chlor-4-nitrobenzoesäure
3-Chloro-4-nitrobenzoic acid
Formel: C₇H₄ClNO₄
MW: 201,57 g/mol
MDL Number: MFCD01632172
CAS-Nummer: 39608-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR