3-Chlor-4-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS25367G1 AS25367G25 AS25367G5
ACTIAS25367G1EA 25 CHF
ACTIAS25367G1 ACTIAS25367G25 ACTIAS25367G5
3-Chlor-4-methoxybenzonitril
3-Chlor-4-methoxybenzonitril
Formel: C₈H₆ClNO
MW: 167,59 g/mol
Siedepunkt: 276 °C (1013 hPa)
Melting Pt: 107…109 °C
MDL Number: MFCD03093073
CAS-Nummer: 102151-33-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR