5-Chlor-2-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS34554G1 AS34554G10 AS34554G5
ACTIAS34554G1EA 0 CHF
ACTIAS34554G1 ACTIAS34554G10 ACTIAS34554G5
5-Chlor-2-methoxybenzonitril
5-Chlor-2-methoxybenzonitril
Formel: C₈H₆ClNO
MW: 167,59 g/mol
Siedepunkt: 276 °C (1013 hPa)
Melting Pt: 99 °C
MDL Number: MFCD00052867
CAS-Nummer: 55877-79-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR