3,5-Dichlor-4-hydroxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS35635G1 AS35635G5
ACTIAS35635G1EA 0 CHF
ACTIAS35635G1 ACTIAS35635G5
3,5-Dichlor-4-hydroxybenzaldehyd
3,5-Dichloro-4-hydroxybenzaldehyde
Formel: C₇H₄Cl₂O₂
MW: 191,01 g/mol
MDL Number: MFCD00017605
CAS-Nummer: 2314-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR