2-Brom-4-methoxyphenol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Brom-4-hydroxyanisole
AS25295G25
AS25295G5
ACTIAS25295G25EA
0
CHF
ACTIAS25295G25
ACTIAS25295G5
2-Brom-4-methoxyphenol
2-Brom-4-methoxyphenol
Formel:
C₇H₇BrO₂ MW: 203.04 g/mol |
MDL Number:
MFCD02113712 CAS-Nummer: 17332-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...