2-Brom-5-methoxyphenol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Brom-3-hydroxyanisole
AS33866G1
AS33866G5
ACTIAS33866G1EA
0
CHF
ACTIAS33866G1
ACTIAS33866G5
2-Brom-5-methoxyphenol
2-Brom-5-methoxyphenol
Formel:
C₇H₇BrO₂ |
MDL Number:
MFCD03428523 CAS-Nummer: 63604-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...