1-Brom-3,5-dinitrobenzol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3,5-Dinitrobromobenzene
AS34587G1
AS34587G5
ACTIAS34587G1EA
0
CHF
ACTIAS34587G1
ACTIAS34587G5
1-Brom-3,5-dinitrobenzol
1-Bromo-3,5-dinitrobenzene
Formel:
C₆H₃BrN₂O₄ MW: 247 g/mol |
MDL Number:
MFCD00156596 CAS-Nummer: 18242-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...