5-Brom-2-nitrophenol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Bromo-3-hydroxy-4-nitrobenzene

AS31808G1 AS31808G5
ACTIAS31808G1EA 0 CHF
ACTIAS31808G1 ACTIAS31808G5
5-Brom-2-nitrophenol
5-Brom-2-nitrophenol
Formel: C₆H₄BrNO₃
MDL Number: MFCD03095027
CAS-Nummer: 27684-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR