5-Brom-3-nitro-2-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-2-methyl-3-nitropyridine

AS32515G1 AS32515G5
ACTIAS32515G1EA 0 CHF
ACTIAS32515G1 ACTIAS32515G5
5-Brom-3-nitro-2-picolin
5-Brom-3-nitro-2-picolin
Formel: C₆H₅BrN₂O₂
MW: 217,02 g/mol
MDL Number: MFCD09031419
CAS-Nummer: 911434-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR