5-Brom-2-fluorphenol

Lieferant: ACTIVATE SCIENTIFIC

AS22832G100 AS22832G25
ACTIAS22832G100EA 0 CHF
ACTIAS22832G100 ACTIAS22832G25
5-Brom-2-fluorphenol
5-Brom-2-fluorphenol
Formel: C₆H₄BrFO
MW: 191 g/mol
MDL Number: MFCD07782066
CAS-Nummer: 112204-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR