4-Brom-2-(trifluormethyl)phenol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Bromo-2-hydroxybenzotrifluoride
AS33829G1
AS33829G10
AS33829G5
ACTIAS33829G1EA
0
CHF
ACTIAS33829G1
ACTIAS33829G10
ACTIAS33829G5
4-Brom-2-(trifluormethyl)phenol
4-Brom-2-(trifluormethyl)phenol
Formel:
C₇H₄BrF₃O MW: 241.01 |
MDL Number:
MFCD05668981 CAS-Nummer: 50824-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...