2-Brom-4-(trifluormethyl)phenol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Bromo-4-hydroxybenzotrifluoride
AS32478G1
AS32478G5
ACTIAS32478G1EA
0
CHF
ACTIAS32478G1
ACTIAS32478G5
2-Brom-4-(trifluormethyl)phenol
2-Brom-4-(trifluormethyl)phenol
Formel:
C₇H₄BrF₃O MW: 241.01 |
MDL Number:
MFCD06657954 CAS-Nummer: 81107-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...