2-Brom-6-fluoranisol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Bromo-3-fluoro-2-methoxybenzene

AS35560G1 AS35560G5
ACTIAS35560G1EA 0 CHF
ACTIAS35560G1 ACTIAS35560G5
2-Brom-6-fluoranisol
2-Brom-6-fluoranisol
Formel: C₇H₆BrFO
MW: 205.03 g/mol
MDL Number: MFCD04974125
CAS-Nummer: 845829-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR