1-Brom-3-fluor-2-nitrobenzol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Bromo-6-fluoronitrobenzene
AS31368G1
AS31368G5
ACTIAS31368G1EA
0
CHF
ACTIAS31368G1
ACTIAS31368G5
1-Brom-3-fluor-2-nitrobenzol
1-Bromo-3-fluoro-2-nitrobenzene
Formel:
C₆H₃BrFNO₂ MW: 220 g/mol |
MDL Number:
MFCD07368788 CAS-Nummer: 886762-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...