1-Brom-2,5-difluor-4-nitrobenzol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1-Brom-2,5-difluor-4-nitrobenzol, 4-Bromo-2,5-difluoronitrobenzene
AS34592G100
AS34592G5
ACTIAS34592G100EA
0
CHF
ACTIAS34592G100
ACTIAS34592G5
1-Brom-2,5-difluor-4-nitrobenzol
1-Bromo-2,5-difluoro-4-nitrobenzene
Formel:
C₆H₂BrF₂NO₂ MW: 237,99 g/mol Melting Pt: 55…57 °C |
MDL Number:
MFCD00042184 CAS-Nummer: 167415-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...