4-Brom-2-fluorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Brom-4-cyan-3-fluorbenzene

AS20100G100 AS20100G25
ACTIAS20100G100EA 0 CHF
ACTIAS20100G100 ACTIAS20100G25
4-Brom-2-fluorbenzonitril
4-Brom-2-fluorbenzonitril
Formel: C₇H₃BrFN
MW: 200,01 g/mol
Siedepunkt: 238…239 °C (1013 hPa)
Melting Pt: 71…73 °C
MDL Number: MFCD00143264
CAS-Nummer: 105942-08-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR