5-Brom-2-fluorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS21013G100 AS21013G25
ACTIAS21013G100EA 232 CHF
ACTIAS21013G100 ACTIAS21013G25
5-Brom-2-fluorbenzonitril
5-Brom-2-fluorbenzonitril
Formel: C₇H₃BrFN
MW: 200,01 g/mol
Siedepunkt: 255 °C (1013 hPa)
Melting Pt: 75…80 °C
MDL Number: MFCD00143424
CAS-Nummer: 179897-89-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR