2-Brom-3-fluorbenzonitril
Lieferant: ACTIVATE SCIENTIFIC
AS24840G25
AS24840G5
ACTIAS24840G25EA
0
CHF
ACTIAS24840G25
ACTIAS24840G5
2-Brom-3-fluorbenzonitril
2-Brom-3-fluorbenzonitril
Formel:
C₇H₃BrFN |
MDL Number:
MFCD07368773 CAS-Nummer: 425379-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...