2-Brom-6-fluorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS24776G100 AS24776G25
ACTIAS24776G100EA 0 CHF
ACTIAS24776G100 ACTIAS24776G25
2-Brom-6-fluorbenzonitril
2-Brom-6-fluorbenzonitril
Formel: C₇H₃BrFN
MDL Number: MFCD06797753
CAS-Nummer: 79544-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR