3-Brom-5-(trifluormethyl)benzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Bromo-5-trifluoromethylbenzaldehyde
AS35176G1
AS35176G5
ACTIAS35176G1EA
0
CHF
ACTIAS35176G1
ACTIAS35176G5
3-Brom-5-(trifluormethyl)benzaldehyd
3-Bromo-5-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄BrF₃O |
MDL Number:
MFCD08059506 CAS-Nummer: 477535-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...